missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Probable dimethyladenosine transferase Protein
A DNA sequence encoding the Homo sapiens (Human) Probable dimethyladenosine transferase, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8253-ec-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001335005.1 | |
Probable dimethyladenosine transferase Protein | |
100 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
27292 | |
Greater than 90% as determined by SDS-PAGE. | |
Research Use Only | |
DIMT1 | |
Recombinant Protein | |
E. coli | |
MPKVKSGAIG RRRGRQEQRR ELKSAGGLMF NTGIGQHILK NPLIINSIID KAALRPTDVV LEVGPGTGNM TVKLLEKAKK VVACELDPRL VAELHKRVQG TPVASKLQVL VGDVLKTDLP FFDTCVANLP YQISSPFVFK LLLHRPFFRC AILMFQREFA LRLVAKPGDK LYCRLSINTQ LLARVDHLMK VGKNNFRPPP KVESSVVRIE PKNPPPPINF QEWDGLVRIT FVRKNKTLSA AFKSSAVQQL LEKNYRIHCS VHNIIIPEDF SIADKIQQIL TSTGFSDKRA RSMDIDDFIR LLHGFNAEGI HFS |