missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Pulmonary surfactant-associated protein A1 Protein
A DNA sequence encoding the Homo sapiens(Human) Pulmonary surfactant-associated protein A1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP10005-iv-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001087239.2 | |
Pulmonary surfactant-associated protein A1 Protein | |
Research Use Only | |
SFTPA1 | |
Recombinant Protein | |
E. coli | |
EVKDVCVGSP GIPGTPGSHG LPGRDGRDGL KGDPGPPGPM GPPGEMPCPP GNDGLPGAPG IPGECGEKGE PGERGPPGLP AHLDEELQAT LHDFRHQILQ TRGALSLQGS IMTVGEKVFS SNGQSITFDA IQEACARAGG RIAVPRNPEE NEAIASFVKK YNTYAYVGLT EGPSPGDFRY SDGTPVNYTN WYRGEPAGRG KEQCVEMYTD GQWNDRNCLY SRLTICEF | |
Greater than 80% as determined by SDS-PAGE. |
653509 | |
100 μg | |
Not determined. | |
Full Length | |
Human | |
His-B2M | |
Tris-based buffer, 50% glycerol |