missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human RAB3A Protein
A cDNA sequence encoding the RAB3A was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13241-1mg
Specifications
RAB3A Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MASATDSRYG QKESSDQNFD YMFKILIIGN SSVGKTSFLF RYADDSFTPA FVSTVGIDFK VKTIYRNDKR IKLQIWDTAG QERYRTITTA YYRGAMGFIL MYDITNEESF NAVQDWSTQI KTYSWDNAQV LLVGNKCDME DERVVSSERG RQLADHLGFE FFEASAKDNI NVKQTFERLV DVICEKMSES LDTADPAVTG AKQGPQLSDQ QVPPHQDCAC | |
Greater than 90.0% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
RAB3A protein solution (0.5 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 20% glycerol, 0.1M NaCl and 1mM DTT. |