missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human RAB3A Protein
A cDNA sequence encoding the RAB3A was constructed and used to recombinantly synthesize the protein.
131.00 € - 7540.00 €
Specifications
Name | RAB3A Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15942949
|
enQuireBio™
QP13241-2UG |
2 μg |
131.00 €
2µg |
Estimated Shipment: 26-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15922949
|
enQuireBio™
QP13241-10UG |
10 μg |
203.00 €
10µg |
Estimated Shipment: 26-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15932949
|
enQuireBio™
QP13241-1MG |
1 mg |
7540.00 €
1mg |
Estimated Shipment: 26-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
RAB3A Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
RAB3A protein solution (0.5 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 20% glycerol, 0.1M NaCl and 1mM DTT. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MASATDSRYG QKESSDQNFD YMFKILIIGN SSVGKTSFLF RYADDSFTPA FVSTVGIDFK VKTIYRNDKR IKLQIWDTAG QERYRTITTA YYRGAMGFIL MYDITNEESF NAVQDWSTQI KTYSWDNAQV LLVGNKCDME DERVVSSERG RQLADHLGFE FFEASAKDNI NVKQTFERLV DVICEKMSES LDTADPAVTG AKQGPQLSDQ QVPPHQDCAC | |
Greater than 90.0% as determined by SDS-PAGE. |