missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human RBP4 Protein
A cDNA sequence encoding the RBP4 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13283-5ug
Additional Details : Weight : 0.01000kg
Specifications
RBP4 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MERDCRVSSF RVKENFDKAR FSGTWYAMAK KDPEGLFLQD NIVAEFSVDE TGQMSATAKGRVRLLNNWDV CADMVGTFTD TEDPAKFKMK YWGVASFLQK GNDDHWIVDT DYDTYAVQYS CRLLNLDGTC ADSYSFVFSR DPNGLPPEAQ KIVRQRQEEL CLARQYRLIV HNGYCDGRSE RNLL | |
Greater than 95.0% as determined by SDS-PAGE. |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
Untagged | |
1 mg/ml in PBS pH 7.4. |