missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human RBP4 Protein
A cDNA sequence encoding the RBP4 was constructed and used to recombinantly synthesize the protein.
131.00 € - 5220.00 €
Specifications
Name | RBP4 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15933079
|
enQuireBio™
QP13283-5UG |
5 μg |
131.00 €
5µg |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15923079
|
enQuireBio™
QP13283-20UG |
20 μg |
203.00 €
20µg |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15913079
|
enQuireBio™
QP13283-1MG |
1 mg |
5220.00 €
1mg |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
RBP4 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
Untagged | |
1 mg/ml in PBS pH 7.4. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MERDCRVSSF RVKENFDKAR FSGTWYAMAK KDPEGLFLQD NIVAEFSVDE TGQMSATAKGRVRLLNNWDV CADMVGTFTD TEDPAKFKMK YWGVASFLQK GNDDHWIVDT DYDTYAVQYS CRLLNLDGTC ADSYSFVFSR DPNGLPPEAQ KIVRQRQEEL CLARQYRLIV HNGYCDGRSE RNLL | |
Greater than 95.0% as determined by SDS-PAGE. |