missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Regulator of microtubule dynamics protein 1
A DNA sequence encoding the Homo sapiens (Human) Regulator of microtubule dynamics protein 3, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7878-ec-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_001291731.1 | |
Regulator of microtubule dynamics protein 1 | |
1 mg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
55177 | |
Greater than 90% as determined by SDS-PAGE. | |
Research Use Only | |
RMDN3 | |
Recombinant Protein | |
E. coli | |
MSRLGALGGA RAGLGLLLGT AAGLGFLCLL YSQRWKRTQR HGRSQSLPNS LDYTQTSDPG RHVMLLRAVP GGAGDASVLP SLPREGQEKV LDRLDFVLTS LVALRREVEE LRSSLRGLAG EIVGEVRCHM EENQRVARRR RFPFVRERSD STGSSSVYFT ASSGATFTDA ESEGGYTTAN AESDNERDSD KESEDGEDEV SCETVKMGRK DSLDLEEEAA SGASSALEAG GSSGLEDVLP LLQQADELHR GDEQGKREGF QLLLNNKLVY GSRQDFLWRL ARAYSDMCEL TEEVSEKKSY ALDGKEEAEA ALEKGDESAD CHLWYAVLCG QLAEHESIQR RIQSGFSFKE HVDKAIALQP ENPMAHFLLG RWCYQVSHLS WLEKKTATAL LESPLSATVE DALQSFLKAE ELQPGFSKAG RVYISKCYRE LGKNSEARWW MKLALELPDV TKEDLAIQKD LEELEVILRD |