missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Rhombotin-1 Protein

A DNA sequence encoding the Homo sapiens (Human) Rhombotin-1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8396-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16044391

  • 518.00 € / 100µg
Estimated Shipment: 04-08-2022
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001257357.1
Rhombotin-1 Protein
100 μg
Not determined.
Partial (see sequence information for more details).
Human
GST
Tris-based buffer, 50% glycerol
4004
Greater than 90% as determined by SDS-PAGE.
Research Use Only
LMO1
Recombinant Protein
E. coli
DKEDGVPMLS VQPKGKQKGC AGCNRKIKDR YLLKALDKYW HEDCLKCACC DCRLGEVGST LYTKANLILC RRDYLRLFGT TGNCAACSKL IPAFEMVMRA RDNVYHLDCF ACQLCNQRFC VGDKFFLKNN MILCQMDYEE GQLNGTFESQ VQ
Videos
SDS
Documents
Certificates
Special Offers

Special Offers