missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human S100B Protein

A cDNA sequence encoding the S100B was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13402-GST-2ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15933459

  • 241.00 € / 2µg
Estimated Shipment: 23-01-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

6285
2 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
MSELEKAMVA LIDVFHQYSG REGDKHKLKK SELKELINNE LSHFLEEIKE QEVVDKVMET LDNDGDGECD FQEFMAFVAM VTTACHEFFE HE
S100B Protein
Research Use Only
S100B
Human
GST
S100B is supplied in 50mM Tris-Acetate, pH 7.5, 1mM EDTA and 20% Glycerol.
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers