missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human S100P Protein

A cDNA sequence encoding the S100P was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13408-HIS-5ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15933479

  • 131.00 € / 5µg
Estimated Shipment: 04-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

S100P Protein
Research Use Only
Recombinant Protein
E. coli
MTELEAAMGM IIDVFSRYSG SEGSTQTLTK GELKVLMEKE LPGFLQSGKD KDAVDKLLKD LDANGDAQVD FSEFIVFVAA ITSACHKYFE KAGLK
Greater than 95% as determined by SDS-PAGE.
5 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
Filtered (0.4microm) and lyophilized from 0.5 mg/ml in 0.05M Phosphate buffer pH 7.2, 0.1M NaCl.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers