missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human S100P Protein
A cDNA sequence encoding the S100P was constructed and used to recombinantly synthesize the protein.
131.00 € - 203.00 €
Specifications
Name | S100P Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15933479
|
enQuireBio™
QP13408-HIS-5UG |
5 μg |
131.00 €
5µg |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15923479
|
enQuireBio™
QP13408-HIS-25UG |
25 μg |
203.00 €
25µg |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
S100P Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
Filtered (0.4microm) and lyophilized from 0.5 mg/ml in 0.05M Phosphate buffer pH 7.2, 0.1M NaCl. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MTELEAAMGM IIDVFSRYSG SEGSTQTLTK GELKVLMEKE LPGFLQSGKD KDAVDKLLKD LDANGDAQVD FSEFIVFVAA ITSACHKYFE KAGLK | |
Greater than 95% as determined by SDS-PAGE. |