missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human SAA Protein

A cDNA sequence encoding the SAA was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP10860-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15945388

  • 131.00 € / 10µg
Estimated Shipment: 26-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

SAA Protein
Research Use Only
The biological activity determined by its ability to down-regulate lipid biosynthesis in aortic smooth muscle cells. The effective concentration was found to be 4microm.
Human
His
Human SAA was lyophilized from a concentrated (1 mg/ml) solution in 20mM Tris-HCl, pH 9.0 and 150mM NaCl.
10 μg
< 1.0 EU per ug protein as determined by the LAL method.
Recombinant Protein
E. coli
RSFFSFLGEA FDGARDMWRA YSDMREANYI GSDKYFHARG NYDAAKRGPG GVWAAEAISN ARENIQRFFG RGAEDSLADQ AANEWGRSGK DPNHFRPAGL PEKY
Greater than 98.0% as determined by: (a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers