missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human SAA Protein
A cDNA sequence encoding the SAA was constructed and used to recombinantly synthesize the protein.
131.00 € - 1065.00 €
Specifications
Name | SAA Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | < 1.0 EU per ug protein as determined by the LAL method. |
Biological Activity | The biological activity determined by its ability to down-regulate lipid biosynthesis in aortic smooth muscle cells. The effective concentration was found to be 4microm. |
Product Type | Recombinant Protein |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15945388
|
enQuireBio™
QP10860-10UG |
10 μg |
131.00 €
10µg |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15965388
|
enQuireBio™
QP10860-50UG |
50 μg |
203.00 €
50µg |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15955388
|
enQuireBio™
QP10860-1MG |
1 mg |
1065.00 €
1mg |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
SAA Protein | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
RSFFSFLGEA FDGARDMWRA YSDMREANYI GSDKYFHARG NYDAAKRGPG GVWAAEAISN ARENIQRFFG RGAEDSLADQ AANEWGRSGK DPNHFRPAGL PEKY | |
Greater than 98.0% as determined by: (a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |
Research Use Only | |
The biological activity determined by its ability to down-regulate lipid biosynthesis in aortic smooth muscle cells. The effective concentration was found to be 4microm. | |
Human | |
His | |
Human SAA was lyophilized from a concentrated (1 mg/ml) solution in 20mM Tris-HCl, pH 9.0 and 150mM NaCl. |