missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Human Sulfatase Modifying Factor 1/SUMF1 His Protein

Product Code. p-7183006 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.05 mg
0.1 mg
Unit Size:
0.05mg
0.1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18245546 0.05 mg 0.05mg
18224076 0.1 mg 0.1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18245546 Supplier Novus Biologicals™ Supplier No. NBP2774880.05MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Highly purified. Generating reliable and reproducible results. Applications: SDS-Page

A recombinant protein with a His-Tag and corresponding to the amino acids 34-374aa of Human Sulfatase Modifying Factor 1/SUMF1.

Source: Baculovirus

Amino Acid Sequence:

SQEAGTGAGA GSLAGSCGCG TPQRPGAHGS SAAAHRYSRE ANAPGPVPGE RQLAHSKMVP IPAGVFTMGT DDPQIKQDGE APARRVTIDA FYMDAYEVSN TEFEKFVNST GYLTEAEKFG DSFVFEGMLS EQVKTNIQQA VAAAPWWLPV KGANWRHPEG PDSTILHRPD HPVLHVSWND AVAYCTWAGK RLPTEAEWEY SCRGGLHNRL FPWGNKLQPK GQHYANIWQG EFPVTNTGED GFQGTAPVDA FPPNGYGLYN IVGNAWEWTS DWWTVHHSVE ETLNPKGPPS GKDRVKKGGS YMCHRSYCYR YRCAARSQNT PDSSASNLGF RCAADRLPTM DHHHHHH>

TRUSTED_SUSTAINABILITY

Specifications

Concentration 0.25 mg/mL
For Use With (Application) SDS-PAGE
Formulation PBS (pH 7.4), 10% glycerol
Gene ID (Entrez) 285362
Molecular Weight (g/mol) 38.1 kDa
Quantity 0.05 mg
Source Baculovirus
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Gene Alias AAPA3037, EC 1.8.99.-, FGEC-alpha-formylglycine-generating enzyme 1, FGly-generating enzyme, MGC131853, MGC150436, sulfatase modifying factor 1, sulfatase-modifying factor 1, UNQ3037
Gene Symbol SUMF1
Buffer PBS (pH 7.4), 10% glycerol.
Endotoxin Level < 1.0 EU per 1 μg of protein (determined by LAL method)
Purity or Quality Grade >85%, by SDS-PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.