missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Suppressor of cytokine signaling 2 Protein

A DNA sequence encoding the Homo sapiens (Human) Suppressor of cytokine signaling 2, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6712-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16055300

  • 3260.00 € / 1mg
Estimated Shipment: 24-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001257396.1
Suppressor of cytokine signaling 2 Protein
Research Use Only
SOCS2
Recombinant Protein
E. coli
MTLRCLEPSG NGGEGTRSQW GTAGSAEEPS PQAARLAKAL RELGQTGWYW GSMTVNEAKE KLKEAPEGTF LIRDSSHSDY LLTISVKTSA GPTNLRIEYQ DGKFRLDSII CVKSKLKQFD SVVHLIDYYV QMCKDKRTGP EAPRNGTVHL YLTKPLYTSA PSLQHLCRLT INKCTGAIWG LPLPTRLKDY LEEYKFQV
Greater than 80% as determined by SDS-PAGE.
8835
1 mg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers