missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human TACI/TNFRSF13B/CVID Protein

Product Code. 18621638 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
1000 μg
5 μg
500 μg
Unit Size:
1000µg
100µg
500µg
5µg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18621638 5 μg 5µg
18608958 100 μg 100µg
18631918 1000 μg 1000µg
18628158 500 μg 500µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 18621638 Supplier R&D Systems™ Supplier No. NBP2613085ug
  • ONLINE EXCLUSIVE PRICE
    199.50 €
    175.35 € / 5µg
    Save 24.15 €
    12% Off
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Novus product, part of R&D Systems. Recombinant Protein

Fully biologically active when compared to standard. The biological activity is determined by its ability to block human BAFF induced T2B cell survival using a concentration range of 1.0-3.0 μg/mL.
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Western Blot, Bioactivity
Formulation Lyophilized from a 0.2 μm filtered concentrated solution in PBS, pH 7.4.
Gene ID (Entrez) 23495
Molecular Weight (g/mol) MolecularWeight-theroretical: 18 kDa
Quantity 5 μg
Storage Requirements Store at -20°C to -70°C as supplied. After reconstitution, store at 2 to 8 C for 1 month and (at -20°C to -70°C for long term storage. Avoid repeated freeze/thaw cycles.)
Endotoxin Concentration Less than 1 EU/μg of TACI/TNFRSF13B/CVID as determined by LAL method.
Gene Alias CD267, CD267 antigen, CVID, CVID2, FLJ39942, MGC133214, TACIMGC39952, TNFRSF14B, Transmembrane activator and CAML interactor, tumor necrosis factor receptor 13 B, tumor necrosis factor receptor superfamily member 13 B, tumor necrosis factor receptor superfamily, member 13 B
Gene Symbol TNFRSF13B
Reconstitution Recommended to centrifuge prior to opening. Reconstitute in sterile distilled water or aqueous buffer containing 0.1% BSA to a concentration of 0.1-1.0 mg/mL Apportion stock solutions into working aliquots and store (at <-20°C.)
Research Category Apoptosis, B Cell Development and Differentiation Markers, Signal Transduction
Product Line Novus
Purity or Quality Grade >95%, by SDS-PAGE and HPLC
Protein TACI/TNFRSF13B/CVID
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.