missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Human Tau 441 Monomer Protein

Product Code. 18022216 Shop All Bio Techne Products
Click to view available options
Quantity:
100 μg
200 μg
500 μg
Unit Size:
1 set
This item is not returnable. View return policy

Product Code. 18022216

Brand: Novus Biologicals™ NBP276795100UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Highly purified and high bioactivity. Generating reliable and reproducible results. Applications: Western Blot, In vitro assay, In vivo assay, SDS-Page

An un-tagged full length Human Recombinant Tau 441 protein Monomer, Accession #: P10636

The Recombinant Human Tau 441 Monomer Protein has been validated for the following applications: Western Blot, In vitro assay, In vivo assay, SDS-Page.
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Western Blot, In vitro Assay, In vivo Assay, SDS-PAGE
Gene ID (Entrez) 4137
Molecular Weight (g/mol) 45.8 kDa
Quantity 100 μg
Source E. coli
Storage Requirements Store at -80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Gene Symbol MAPT
Specific Reactivity Tau 441
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Buffer 10 mM HEPES, 100 mM NaCl pH 7.4
Purity or Quality Grade 0.2 Micrometer Filtered
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.