missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human TBC1D13 Protein

A cDNA sequence encoding the TBC1D13 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13693-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15974399

  • 3915.00 € / 1mg
Estimated Shipment: 26-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

TBC1D13 Protein
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMSSLHKS RIADFQDVLK EPSIALEKLR ELSFSGIPCE GGLRCLCWKI LLNYLPLERA SWTSILAKQR ELYAQFLREM IIQPGIAKAN MGVSREDVTF EDHPLNPNPD SRWNTYFKDN EVLLQIDKDV RRLCPDISFF QRATDYPCLL ILDPQNEFET LRKRVEQTTL KSQTVARNRS GVTNMSSPHK NSVPSSLNEY EVLPNGCEAH WEVVERILFI YAKLNPGIAY VQGMNEIVGP LYYTFATDPN SEWKEHAEAD TFFCFTNLMA EIRDNFIKSL DDSQCGITYK MEKVYSTLKD KDVELYLKLQ EQNIKPQFFA FRWLTLLLSQ EFLLPDVIRI WDSLFADDNR FDFLLLVCCA MLMLIREQLL EGDFTVNMRL LQDYPITDVC QILQKAKELQ DSK
Greater than 95.0% as determined by SDS-PAGE.
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The TBC1D13 solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 10% glycerol and 1mM DTT.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers