missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human TBC1D13 Protein
A cDNA sequence encoding the TBC1D13 was constructed and used to recombinantly synthesize the protein.
131.00 € - 3915.00 €
Specifications
Name | TBC1D13 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15994399
|
enQuireBio™
QP13693-5UG |
5 μg |
131.00 €
5µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15984399
|
enQuireBio™
QP13693-20UG |
20 μg |
203.00 €
20µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15974399
|
enQuireBio™
QP13693-1MG |
1 mg |
3915.00 €
1mg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
TBC1D13 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The TBC1D13 solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 10% glycerol and 1mM DTT. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMSSLHKS RIADFQDVLK EPSIALEKLR ELSFSGIPCE GGLRCLCWKI LLNYLPLERA SWTSILAKQR ELYAQFLREM IIQPGIAKAN MGVSREDVTF EDHPLNPNPD SRWNTYFKDN EVLLQIDKDV RRLCPDISFF QRATDYPCLL ILDPQNEFET LRKRVEQTTL KSQTVARNRS GVTNMSSPHK NSVPSSLNEY EVLPNGCEAH WEVVERILFI YAKLNPGIAY VQGMNEIVGP LYYTFATDPN SEWKEHAEAD TFFCFTNLMA EIRDNFIKSL DDSQCGITYK MEKVYSTLKD KDVELYLKLQ EQNIKPQFFA FRWLTLLLSQ EFLLPDVIRI WDSLFADDNR FDFLLLVCCA MLMLIREQLL EGDFTVNMRL LQDYPITDVC QILQKAKELQ DSK | |
Greater than 95.0% as determined by SDS-PAGE. |