missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Tegument protein VP16 Protein
A DNA sequence encoding the Homo sapiens (Human) Tegument protein VP16, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7282-ec-100ug
Additional Details : Weight : 0.01000kg
Specifications
Tegument protein VP16 Protein | |
100 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
Greater than 90% as determined by SDS-PAGE. | |
Research Use Only | |
UL48 | |
Recombinant Protein | |
E. coli | |
MDLLVDELFA DMDADGASPP PPRPAGGPKN TPAAPPLYAT GRLSQAQLMP SPPMPVPPAA LFNRLLDDLG FSAGPALCTM LDTWNEDLFS ALPTNADLYR ECKFLSTLPS DVVEWGDAYV PERAQIDIRA HGDVAFPTLP ATRDGLGLYY EALSRFFHAE LRAREESYRT VLANFCSALY RYLRASVRQL HRQAHMRGRD RDLGEMLRAT IADRYYRETA RLARVLFLHL YLFLTREILW AAYAEQMMRP DLFDCLCCDL ESWRQLAGLF QPFMFVNGAL TVRGVPIEAR RLRELNHIRE HLNLPLVRSA ATEEPGAPLT TPPTLHGNQA RASGYFMVLI RAKLDSYSSF TTSPSEAVMR EHAYSRARTK NNYGSTIEGL LDLPDDDAPE EAGLAAPRLS FLPAGHTRRL STAPPTDVSL GDELHLDGED VAMAHADALD DFDLDMLGDG DSPGPGFTPH DSAPYGALDM ADFEFEQMFT DALGIDEYGG |