missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human THAP domain-containing protein 1

A DNA sequence encoding the Homo sapiens (Human) THAP domain-containing protein 6, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7886-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16031401

  • 505.00 € / 100µg
Estimated Shipment: 25-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001304720.1
THAP domain-containing protein 1
Research Use Only
THAP6
Recombinant Protein
E. coli
MVKCCSAIGC ASRCLPNSKL KGLTFHVFPT DENIKRKWVL AMKRLDVNAA GIWEPKKGDV LCSRHFKKTD FDRSAPNIKL KPGVIPSIFD SPYHLQGKRE KLHCRKNFTL KTVPATNYNH HLVGASSCIE EFQSQFIFEH SYSVMDSPKK LKHKLDHVIG ELEDTKESLR NVLDREKRFQ KSLRKTIREL KDECLISQET ANRLDTFCWD CCQESIEQDY IS
Greater than 90% as determined by SDS-PAGE.
152815
100 μg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers