missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human THAP domain-containing protein 1
A DNA sequence encoding the Homo sapiens (Human) THAP domain-containing protein 6, was expressed in the hosts and tags indicated.
160.00 € - 2285.00 €
Specifications
Accession Number | NP_001304720.1 |
---|---|
Gene ID (Entrez) | 152815 |
Name | THAP domain-containing protein 1 |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16031401
|
enQuireBio™
QP7886-EC-100UG |
100 μg |
505.00 €
100µg |
Estimated Shipment: 26-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16051401
|
enQuireBio™
QP7886-EC-1MG |
1 mg |
2285.00 €
1mg |
Estimated Shipment: 26-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16041401
|
enQuireBio™
QP7886-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16061401
|
enQuireBio™
QP7886-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16071401
|
enQuireBio™
QP7886-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16081401
|
enQuireBio™
QP7886-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001304720.1 | |
THAP domain-containing protein 1 | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
152815 | |
Research Use Only | |
THAP6 | |
Recombinant Protein | |
E. coli | |
MVKCCSAIGC ASRCLPNSKL KGLTFHVFPT DENIKRKWVL AMKRLDVNAA GIWEPKKGDV LCSRHFKKTD FDRSAPNIKL KPGVIPSIFD SPYHLQGKRE KLHCRKNFTL KTVPATNYNH HLVGASSCIE EFQSQFIFEH SYSVMDSPKK LKHKLDHVIG ELEDTKESLR NVLDREKRFQ KSLRKTIREL KDECLISQET ANRLDTFCWD CCQESIEQDY IS | |
Greater than 90% as determined by SDS-PAGE. |