missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human TRH Protein
A cDNA sequence encoding the TRH was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13814-100mg
Additional Details : Weight : 0.01000kg
Specifications
TRH Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
Pyr-His-Pro-NH2 | |
Greater than 98.5% as determined by Analysis by RP-HPLC. |
100 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
Untagged | |
The TRH was lyophilized with no additives. |