missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human TRH Protein
A cDNA sequence encoding the TRH was constructed and used to recombinantly synthesize the protein.
373.00 € - 1100.00 €
Specifications
Name | TRH Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15954789
|
enQuireBio™
QP13814-100MG |
100 mg |
373.00 €
100mg |
Estimated Shipment: 28-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15964789
|
enQuireBio™
QP13814-250MG |
250 mg |
725.00 €
250mg |
Estimated Shipment: 28-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15974789
|
enQuireBio™
QP13814-500MG |
500 mg |
1100.00 €
500mg |
Estimated Shipment: 28-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
TRH Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
Untagged | |
The TRH was lyophilized with no additives. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
Pyr-His-Pro-NH2 | |
Greater than 98.5% as determined by Analysis by RP-HPLC. |