missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human TRPA1 Protein

A DNA sequence encoding the Homo sapiens (Human) TRPA1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6843-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16036230

  • 735.00 € / 100µg
Estimated Shipment: 25-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_015628.2
TRPA1 Protein
Research Use Only
TRPA1
Recombinant Protein
E. coli
IGLAVGDIAE VQKHASLKRI AMQVELHTSL EKKLPLWFLR KVDQKSTIVY PNKPRSGGML FHIFCFLFCT GEIRQEIPNA DKSLEMEILK QKYRLKDLTF LLEKQHELIK LIIQKMEIIS ETEDDDSHCS FQDRFKKEQM EQRNSRWNTV LRAVKAKTHH LEP
Greater than 90% as determined by SDS-PAGE.
8989
100 μg
Not determined.
Partial (see sequence information for more details).
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers