missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human TRPA1 Protein
A DNA sequence encoding the Homo sapiens (Human) TRPA1, was expressed in the hosts and tags indicated.
245.00 € - 3260.00 €
Specifications
Accession Number | NP_015628.2 |
---|---|
Gene ID (Entrez) | 8989 |
Name | TRPA1 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16036230
|
enQuireBio™
QP6843-EC-100UG |
100 μg |
735.00 €
100µg |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16056230
|
enQuireBio™
QP6843-EC-1MG |
1 mg |
3260.00 €
1mg |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16046230
|
enQuireBio™
QP6843-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16066230
|
enQuireBio™
QP6843-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16076230
|
enQuireBio™
QP6843-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16086230
|
enQuireBio™
QP6843-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_015628.2 | |
TRPA1 Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
8989 | |
Research Use Only | |
TRPA1 | |
Recombinant Protein | |
E. coli | |
IGLAVGDIAE VQKHASLKRI AMQVELHTSL EKKLPLWFLR KVDQKSTIVY PNKPRSGGML FHIFCFLFCT GEIRQEIPNA DKSLEMEILK QKYRLKDLTF LLEKQHELIK LIIQKMEIIS ETEDDDSHCS FQDRFKKEQM EQRNSRWNTV LRAVKAKTHH LEP | |
Greater than 90% as determined by SDS-PAGE. |