missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Human TWEAK R/TNFRSF12 Protein

Product Code. 18400172 Shop All Bio Techne Products
Click to view available options
Quantity:
1 mg
100 μg
5 μg
500 μg
Unit Size:
100µg
1mg
500µg
5µg
This item is not returnable. View return policy

Product Code. 18400172

Brand: Novus Biologicals™ NBP235085100ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

TWEAK R/TNFRSF12 protein is fully biologically active when compared to standard. The ED50 as determined by inhibiting TWEAK-dependent proliferation of human umbilical vein endothelial cells (HUVEC) is less than 30 ng/mL, corresponding to a specific activity of >3.3 x 104 IU/mg, in the presence of 15 ng/mL of rHuTWEAK
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9NP84, Q9NP84, Q9NP84
Concentration LYOPH
For Use With (Application) PAGE, Bioactivity
Formulation Lyophilized from a 0.2 um filtered concentrated solution in PBS, pH 7.4.
Gene ID (Entrez) 51330
Molecular Weight (g/mol) 5.6 kDa
Quantity 100 μg
Storage Requirements Store at -20°C to -70°C as supplied. After reconstitution, store at 2°C to 8°C for 1 month and at -20°C to -70°C for long term storage. Avoid repeated freeze-thaw cycles.
Endotoxin Concentration Less than 1 EU/ug of TWEAK R/TNFRSF12 as determined by LAL method.
Gene Alias CD266, CD266 antigen, FGF-inducible 14, Fibroblast growth factor-inducible immediate-early response protein 14, FN14tumor necrosis factor receptor superfamily member 12A, tumor necrosis factor receptor superfamily, member 12A, TWEAK Receptor/TNFRSF12A, TweakR, tweak-receptor, type I transmembrane protein Fn14
Gene Symbol TNFRSF12A
Reconstitution Recommended to centrifuge prior to opening. Reconstitute in sterile distilled water or aqueous buffer containing 0.1% BSA to a concentration of 0.1-1.0mg/mL. Apportion stock solutions into working aliquots and store at <-20°C.
Research Category Biologically Active Proteins
Purity or Quality Grade >95% by SDS-PAGE and HPLC
Protein TWEAK R/TNFRSF12
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.