missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Human TWEAK R/TNFRSF12 Protein
Shop All Bio Techne Products
Click to view available options
Quantity:
1 mg
100 μg
5 μg
500 μg
Unit Size:
100µg
1mg
500µg
5µg
Description
TWEAK R/TNFRSF12 protein is fully biologically active when compared to standard. The ED50 as determined by inhibiting TWEAK-dependent proliferation of human umbilical vein endothelial cells (HUVEC) is less than 30 ng/mL, corresponding to a specific activity of >3.3 x 104 IU/mg, in the presence of 15 ng/mL of rHuTWEAK
Specifications
Specifications
| Accession Number | Q9NP84, Q9NP84, Q9NP84 |
| Concentration | LYOPH |
| For Use With (Application) | PAGE, Bioactivity |
| Formulation | Lyophilized from a 0.2 um filtered concentrated solution in PBS, pH 7.4. |
| Gene ID (Entrez) | 51330 |
| Molecular Weight (g/mol) | 5.6 kDa |
| Quantity | 100 μg |
| Storage Requirements | Store at -20°C to -70°C as supplied. After reconstitution, store at 2°C to 8°C for 1 month and at -20°C to -70°C for long term storage. Avoid repeated freeze-thaw cycles. |
| Endotoxin Concentration | Less than 1 EU/ug of TWEAK R/TNFRSF12 as determined by LAL method. |
| Gene Alias | CD266, CD266 antigen, FGF-inducible 14, Fibroblast growth factor-inducible immediate-early response protein 14, FN14tumor necrosis factor receptor superfamily member 12A, tumor necrosis factor receptor superfamily, member 12A, TWEAK Receptor/TNFRSF12A, TweakR, tweak-receptor, type I transmembrane protein Fn14 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction