missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-533 of Human USH1C The Recombinant Human USH1C Protein is derived from E. coli. The Recombinant Human USH1C Protein has been validated for the following applications: SDS-Page.
Specifications
Specifications
| Accession Number | AAH16057 |
| For Use With (Application) | ELISA |
| Formulation | 20mM Tris-HCl buffer (pH 8.0), 20% glycerol |
| Gene ID (Entrez) | 10083 |
| Molecular Weight (g/mol) | 64.6kDa |
| Name | USH1C Protein |
| Purification Method | Protein |
| Immunogen | MRGSHHHHHH GMASMTGGQQ MGRDLYDDDD KDRWGSHMDR KVAREFRHKV DFLIENDAEK DYLYDVLRMY HQTMDVAVLV GDLKLVINEP SRLPLFDAIR PLIPLKHQVE YDQLTPRRSR KLKEVRLDRL HPEGLGLSVR GGLEFGCGLF ISHLIKGGQA DSVGLQVGDE IVRINGYSIS SCTHEEVINL IRTKKTVSIK VRHIGLIPVK SSPDEPLTWQ YVDQFVSESG GVRGSLGSPG NRENKEKKVF ISLVGSRGLG CSISSGPIQK PGIFISHVKP GSLSAEVGLE IGDQIVEVNG VDFSNLDHKE GRELFMTDRE RLAEARQREL QRQELLMQKR LAMESNKILQ EQQEMERQRR KEIAQKAAEE NERYRKEMEQ IVEEEEKFKK QWEEDWGSKE QLLLPKTITA EVHPVPLRKP KYDQGVEPEL EPADDLDGGT EEQGEQDFRK YEEGFDPYSM FTPEQIMGKD VRLLRIKKEG SLDLALEGGV DSPIGKVVVS AVYERGAAER HGGIVKGDEI MAINGKIVTD YTLAEADAAL QKAWNQGGDW IDLVVAVCPP KEYDDELTFF |
| Storage Requirements | -80°C. Avoid freeze-thaw cycles. |
| Cross Reactivity | Human |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?