missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Vasculin Protein

A DNA sequence encoding the Homo sapiens (Human) Vasculin, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7795-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16050841

  • 505.00 € / 100µg
Estimated Shipment: 05-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001120707.2
Vasculin Protein
Research Use Only
GPBP1
Recombinant Protein
E. coli
MRTDKKSEFL KALKRDRVEE EHEDESRAGS EKDDDSFNLH NSNSTHQERD INRNFDENEI PQENGNASVI SQQIIRSSTF PQTDVLSSSL EAEHRLLKEM GWQEDSENDE TCAPLTEDEM REFQVISEQL QKNGLRKNGI LKNGLICDFK FGPWKNSTFK PTTENDDTET SSSDTSDDDD V
Greater than 90% as determined by SDS-PAGE.
65056
100 μg
Not determined.
Partial (see sequence information for more details).
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers