missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human VEGF-121 Isoform Protein
Recombinant Protein
Brand: enQuireBio™ QP5207-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001165099.1 | |
500 μg | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
HUVEC proliferation was significantly induced by less than 5 ng/ml of Human VEGF-121 Isoform, or activity of at least 2.0 x 10^5 units/mg. | |
Human | |
Untagged | |
Sterile filtered (0.22 um) solution containing 0.1% TFA |
VEGF-121 Isoform Protein | |
Research Use Only | |
VEGFA | |
Recombinant Protein | |
E. coli | |
MAPMAEGGGQ NHHEVVKFMD VYQRSYCHPI ETLVDIFQEY PDEIEYIFKP SCVPLMRCGG CCNDEGLECV PTEESNITMQ IMRIKPHQGQ HIGEMSFLQH NKCECRPKKD RARQENCDKP RR | |
Greater than 95% as determined by SDS-PAGE. |