missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human VEGF-121 Isoform Protein
Recombinant Protein
107.00 € - 5365.00 €
Specifications
Accession Number | NP_001165099.1 |
---|---|
Name | VEGF-121 Isoform Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Less than or equal to 1.00 EUs/ug as determined by the LAL method. |
Gene Symbol | VEGFA |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15965679
|
enQuireBio™
QP5207-2UG |
2 μg |
107.00 €
2µg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15945679
|
enQuireBio™
QP5207-10UG |
10 μg |
252.00 €
10µg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15935679
|
enQuireBio™
QP5207-100UG |
100 μg |
700.00 €
100µg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15975679
|
enQuireBio™
QP5207-500UG |
500 μg |
3285.00 €
500µg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15955679
|
enQuireBio™
QP5207-1MG |
1 mg |
5365.00 €
1mg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
NP_001165099.1 | |
Research Use Only | |
VEGFA | |
Recombinant Protein | |
E. coli | |
MAPMAEGGGQ NHHEVVKFMD VYQRSYCHPI ETLVDIFQEY PDEIEYIFKP SCVPLMRCGG CCNDEGLECV PTEESNITMQ IMRIKPHQGQ HIGEMSFLQH NKCECRPKKD RARQENCDKP RR | |
Greater than 95% as determined by SDS-PAGE. |
VEGF-121 Isoform Protein | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
HUVEC proliferation was significantly induced by less than 5 ng/ml of Human VEGF-121 Isoform, or activity of at least 2.0 x 10^5 units/mg. | |
Human | |
Untagged | |
Sterile filtered (0.22 um) solution containing 0.1% TFA |