missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ZFAND5 Protein
A cDNA sequence encoding the ZFAND5 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13994-20ug
Additional Details : Weight : 0.01000kg
Specifications
ZFAND5 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMAQETNQ TPGPMLCSTG CGFYGNPRTN GMCSVCYKEH LQRQQNSGRM SPMGTASGSN SPTSDSASVQ RADTSLNNCE GAAGSTSEKS RNVPVAALPV TQQMTEMSIS REDKITTPKT EVSEPVVTQP SPSVSQPSTS QSEEKAPELP KPKKNRCFMC RKKVGLTGFD CRCGNLFCGL HRYSDKHNCP YDYKAEAAAK IRKENPVVVA EKIQRI | |
Greater than 85.0% as determined by SDS-PAGE. |
20 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
ZFAND5 protein solution (0.5 mg/ml) containing 20mM Phosphate buffer saline (pH 8.0), 30% glycerol and 1mM DTT. |