missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ZFAND5 Protein
A cDNA sequence encoding the ZFAND5 was constructed and used to recombinantly synthesize the protein.
131.00 € - 3915.00 €
Specifications
Name | ZFAND5 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15995349
|
enQuireBio™
QP13994-5UG |
5 μg |
131.00 €
5µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15985349
|
enQuireBio™
QP13994-20UG |
20 μg |
203.00 €
20µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15975349
|
enQuireBio™
QP13994-1MG |
1 mg |
3915.00 €
1mg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
ZFAND5 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
ZFAND5 protein solution (0.5 mg/ml) containing 20mM Phosphate buffer saline (pH 8.0), 30% glycerol and 1mM DTT. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMAQETNQ TPGPMLCSTG CGFYGNPRTN GMCSVCYKEH LQRQQNSGRM SPMGTASGSN SPTSDSASVQ RADTSLNNCE GAAGSTSEKS RNVPVAALPV TQQMTEMSIS REDKITTPKT EVSEPVVTQP SPSVSQPSTS QSEEKAPELP KPKKNRCFMC RKKVGLTGFD CRCGNLFCGL HRYSDKHNCP YDYKAEAAAK IRKENPVVVA EKIQRI | |
Greater than 85.0% as determined by SDS-PAGE. |