missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ZRANB2 Protein
A DNA sequence encoding the Homo sapiens (Human) ZRANB2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6912-ec-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_005446.2 | |
ZRANB2 Protein | |
Research Use Only | |
ZRANB2 | |
Recombinant Protein | |
E. coli | |
MSTKNFRVSD GDWICPDKKC GNVNFARRTS CNRCGREKTT EAKMMKAGGT EIGKTLAEKS RGLFSANDWQ CKTCSNVNWA RRSECNMCNT PKYAKLEERT GYGGGFNERE NVEYIEREES DGEYDEFGRK KKKYRGKAVG PASILKEVED KESEGEEEDE DEDLSKYKLD EDEDEDDADL SKYNLDASEE EDSNKKKSNR RSRSKSRSSH SRSSSRSSSP SSSRSRSRSR SRSSSSSQSR SRSSSRERSR SRGSKSRSSS RSHRGSSSPR KRSYSSSSSS PERNRKRSRS RSSSSGDRKK RRTRSRSPES QVIGENTKQP | |
Greater than 90% as determined by SDS-PAGE. |
9406 | |
100 μg | |
Not determined. | |
Full Length of Isoform 2 | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |