Learn More
enQuireBio™ Recombinant Influenza A H1N1 (Fort Warren / 1 / 1950) Nucleocapsid Protein
A DNA sequence encoding the Influenza A virus (strain A/Fort Warren/1/1950 H1N1) Influenza A H1N1 (strain A/Fort Warren/1/1950) Nucleocapsid, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7081-100ug
Additional Details : Weight : 0.01000kg
Specifications
Influenza A H1N1 (Fort Warren / 1 / 1950) Nucleocapsid Protein | |
100 μg | |
Not determined. | |
Full Length | |
Influenza | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
Greater than 90% as determined by SDS-PAGE. | |
Research Use Only | |
NP | |
Recombinant Protein | |
E. coli | |
MASQGTKRSY EQMETDGDRQ NATEIRASVG KMIDGIGRFY IQMCTELKLS DYEGRLIQNS LTIERMVLSA FDERRNKYLE EHPSAGKDPK KTGGPIYKRV DGKWMRELVL YDKEEIRRIW RQANNGDDAT AGLTHMMIWH SNLNDTTYQR TRALVRTGMD PRMCSLMQGS TLPRRSGAAG AAVKGVGTMV MELIRMIKRG INDRNFWRGE NGRKTRIAYE RMCNILKGKF QTAAQRAMMD QVRESRNPGN AEIEDLIFLA RSALILRGSV AHKSCLPACV YGPAVASGYD FEKEGYSLVG IDPFKLLQNS QVYSLIRPNE NPAHKSQLVW MACNSAAFED LRVSSFIRGT KVIPRGKLST RGVQIASNEN MDTMGSSTLE LRSRYWAIRT RSGGNTNQQR ASAGQISIQP TFSVQRNLPF DKTTIMAAFT GNAEGRTSDM RAEIIRMMES ARPEEVSFQG RGVFELSDER AANPIVPSFD MSNEGSYFFG DNAEEYDN |