missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Influenza A H1N1 (New Jersey / 8 / 1976) NucleoProtein

A DNA sequence encoding the Influenza A virus Influenza A H1N1 (strain A/New Jersey/8/1976) Nucleoprotein, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7117-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16037420

  • 3590.00 € / 1mg
Estimated Shipment: 28-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

Influenza A H1N1 (New Jersey / 8 / 1976) NucleoProtein
Research Use Only
NP
Recombinant Protein
E. coli
MASQGTKRSY EQMETGGERQ DATEIRASVG RMIGGIGRFY IQMCTELKLS DYEGRLIQNS ITIERMVLSA FDERRNKYLE EHPSAGKDPK KTGGPIYKRI DGKWMRELIL YDKEEIRRVW RQANNGEDAT AGLTHIMIWH SNLNDATYQR TRALVRTGMD PRMCSLMQGS TLPRRSGAAG AAVKGVGTIA MELIGMIKRG INDRNFWRGE NGRRTRIAYE RMCNILKGKF QTAAQRAMMD QVRESRNPGN AEIEDLIFLA RSALILRGSV AHKSCLPACV YGLAVASGHD FEREGYSLVG IDPFKLLQNS QVFSLIRPNE NPAHKSQLVW MACHSAAFED LRVSGFIRGK KVVPRGRLST RGVQIASNEN VEAMDSSTLE LRSRYWAIRT RSGGNTNQQK ASADQISVQP TFSVQRNLPF ERATVMAAFI GDNEGRTSDM RTEIIRMMES AKPEDLSFQG RGVFELSDEK ATNPIVPSFD MNNEGSYFFG DNAEEYDN
Greater than 90% as determined by SDS-PAGE.
1 mg
Not determined.
Full Length
Influenza
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers