missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Influenza A H1N1 (New Jersey / 8 / 1976) NucleoProtein
A DNA sequence encoding the Influenza A virus Influenza A H1N1 (strain A/New Jersey/8/1976) Nucleoprotein, was expressed in the hosts and tags indicated.
421.00 € - 3590.00 €
Specifications
Name | Influenza A H1N1 (New Jersey / 8 / 1976) NucleoProtein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Gene Symbol | NP |
Protein Length | Full Length |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16027420
|
enQuireBio™
QP7117-10UG |
10 μg |
421.00 €
10µg |
Estimated Shipment: 28-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16067420
|
enQuireBio™
QP7117-50UG |
50 μg |
605.00 €
50µg |
Estimated Shipment: 28-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16017420
|
enQuireBio™
QP7117-100UG |
100 μg |
1025.00 €
100µg |
Estimated Shipment: 28-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16047420
|
enQuireBio™
QP7117-200UG |
200 μg |
1690.00 €
200µg |
Estimated Shipment: 28-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16057420
|
enQuireBio™
QP7117-500UG |
500 μg |
2260.00 €
500µg |
Estimated Shipment: 28-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16037420
|
enQuireBio™
QP7117-1MG |
1 mg |
3590.00 €
1mg |
Estimated Shipment: 28-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
Influenza A H1N1 (New Jersey / 8 / 1976) NucleoProtein | |
Not determined. | |
Full Length | |
Influenza | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
Research Use Only | |
NP | |
Recombinant Protein | |
E. coli | |
MASQGTKRSY EQMETGGERQ DATEIRASVG RMIGGIGRFY IQMCTELKLS DYEGRLIQNS ITIERMVLSA FDERRNKYLE EHPSAGKDPK KTGGPIYKRI DGKWMRELIL YDKEEIRRVW RQANNGEDAT AGLTHIMIWH SNLNDATYQR TRALVRTGMD PRMCSLMQGS TLPRRSGAAG AAVKGVGTIA MELIGMIKRG INDRNFWRGE NGRRTRIAYE RMCNILKGKF QTAAQRAMMD QVRESRNPGN AEIEDLIFLA RSALILRGSV AHKSCLPACV YGLAVASGHD FEREGYSLVG IDPFKLLQNS QVFSLIRPNE NPAHKSQLVW MACHSAAFED LRVSGFIRGK KVVPRGRLST RGVQIASNEN VEAMDSSTLE LRSRYWAIRT RSGGNTNQQK ASADQISVQP TFSVQRNLPF ERATVMAAFI GDNEGRTSDM RTEIIRMMES AKPEDLSFQG RGVFELSDEK ATNPIVPSFD MNNEGSYFFG DNAEEYDN | |
Greater than 90% as determined by SDS-PAGE. |