Learn More
enQuireBio™ Recombinant Influenza A H1N1 (New Jersey / 8 / 1976) NucleoProtein
A DNA sequence encoding the Influenza A virus Influenza A H1N1 (strain A/New Jersey/8/1976) Nucleoprotein, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7117-200ug
Additional Details : Weight : 0.01000kg
Specifications
Influenza A H1N1 (New Jersey / 8 / 1976) NucleoProtein | |
200 μg | |
Not determined. | |
Full Length | |
Influenza | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
Greater than 90% as determined by SDS-PAGE. | |
Research Use Only | |
NP | |
Recombinant Protein | |
E. coli | |
MASQGTKRSY EQMETGGERQ DATEIRASVG RMIGGIGRFY IQMCTELKLS DYEGRLIQNS ITIERMVLSA FDERRNKYLE EHPSAGKDPK KTGGPIYKRI DGKWMRELIL YDKEEIRRVW RQANNGEDAT AGLTHIMIWH SNLNDATYQR TRALVRTGMD PRMCSLMQGS TLPRRSGAAG AAVKGVGTIA MELIGMIKRG INDRNFWRGE NGRRTRIAYE RMCNILKGKF QTAAQRAMMD QVRESRNPGN AEIEDLIFLA RSALILRGSV AHKSCLPACV YGLAVASGHD FEREGYSLVG IDPFKLLQNS QVFSLIRPNE NPAHKSQLVW MACHSAAFED LRVSGFIRGK KVVPRGRLST RGVQIASNEN VEAMDSSTLE LRSRYWAIRT RSGGNTNQQK ASADQISVQP TFSVQRNLPF ERATVMAAFI GDNEGRTSDM RTEIIRMMES AKPEDLSFQG RGVFELSDEK ATNPIVPSFD MNNEGSYFFG DNAEEYDN |