missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Influenza H3N2 Kiev Protein

Recombinant Protein

Brand:  enQuireBio™ QP12083-50ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15929318

  • 203.00 € / 50µg
Estimated Shipment: 27-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

Influenza H3N2 Kiev Protein
Research Use Only
Recombinant Protein
Baculovirus
The H3N2 A/Kiev/301/94 solution contains 0.1M NaCl, 10mM Tris-Hcl, 1mM EDTA pH 8.0, 0.1% sodium azide (NaN3) and 0.005% thimerosal.
50 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Influenza
Untagged
Greater than 90.0% as determined byAnalysis by SDS-PAGE.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers