missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Influenza H3N2 Protein
A cDNA sequence encoding the H3N2 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12080-20ug
Additional Details : Weight : 0.01000kg
Specifications
Influenza H3N2 Protein | |
Research Use Only | |
Recombinant Protein | |
Baculovirus | |
ADNLPGNENN AATLCLGHHA VPNGTIVKTI TDDQIEVTNA TELVQNSSTG KICNNPHKIL DGRDCTLIDA LLGDPHCDVF QNETWDLFVE RSNAFSNCYP YDVPDYASLR SIVASSGTLE FITEGFTWAG VTQNGGSGAC KKGPANGFFS RLNWLTKSGN TYPVLNVTMP NNNNFDKLYI WGVHHPSTNQ EQTSLYIQAS GRVKVSTRRS QQTIIPNIGS RPLVRGQSGR ISVYWTIVKP GDVLVINSNG NLIAPRGYFK MRIGKSSIMR SDAPIDTCIS ECITPNGSIP NEKPFQNVNK ITYGACPKYV KQNTLKLATG MRNVPERQTH HHHHH | |
Greater than 90% as determined by SDS-PAGE. |
20 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Influenza | |
His | |
The H3N2 Canine solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0) and 10% glycerol. |