missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Macaque IL17 / IL17A Protein
A DNA sequence encoding the Macaca mulatta (Rhesus macaque) IL17 / IL17A, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6211-ec-100ug
Additional Details : Weight : 0.01000kg
Specifications
XP_001106391.1 | |
IL17 / IL17A Protein | |
100 μg | |
Not determined. | |
Full Length | |
Macaque | |
GST | |
Tris-based buffer, 50% glycerol |
708123 | |
Greater than 90% as determined by SDS-PAGE. | |
Research Use Only | |
IL17A | |
Recombinant Protein | |
E. coli | |
GIAIPRNPGC PNSEDKTFPR TVMVNLNIHN RNTNTNPKRS SDYYNRSTSP WNLHRNEDPE RYPSVIWEAK CRHLGCVNAD GNVDYHMNSV PIQQEILVLR REPRHCPNSF RLEKILVSVG CTCVTPIVHH VA |