missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Measles Virus MV Large Polymerase 58-149 a. a Protein
A cDNA sequence encoding the MV Large Polymerase 58-149 a. a was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12758-1mg
Additional Details : Weight : 0.01000kg
Specifications
MV Large Polymerase 58-149 a. a Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
50mM Tris-HCl, pH 8.0, 60mM NaCl, 10mM glutathione, 0.1% sarcosil and 50% glycerol. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Measles Virus | |
Untagged | |
Measles Large Polymerase is >95% pure as determined by 10% PAGE (coomassie staining) and RP-HPLC. |