missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Measles Virus MV Large Polymerase 58-149 a. a Protein
A cDNA sequence encoding the MV Large Polymerase 58-149 a. a was constructed and used to recombinantly synthesize the protein.
228.00 € - 1740.00 €
Specifications
Name | MV Large Polymerase 58-149 a. a Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Measles Virus |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15951449
|
enQuireBio™
QP12758-100UG |
100 μg |
228.00 €
100µg |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15971449
|
enQuireBio™
QP12758-500UG |
500 μg |
870.00 €
500µg |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15961449
|
enQuireBio™
QP12758-1MG |
1 mg |
1740.00 €
1mg |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Spezifikation
MV Large Polymerase 58-149 a. a Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Measles Virus | |
Untagged | |
Measles Large Polymerase is >95% pure as determined by 10% PAGE (coomassie staining) and RP-HPLC. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
50mM Tris-HCl, pH 8.0, 60mM NaCl, 10mM glutathione, 0.1% sarcosil and 50% glycerol. |