missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse aFGF / FGF1 Protein

A DNA sequence encoding the Mus musculus (Mouse) aFGF / FGF1 Protein, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6037-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15959529

  • 3260.00 € / 1mg
Estimated Shipment: 10-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_034327.1
aFGF / FGF1 Protein
Research Use Only
FGF1
Recombinant Protein
E. coli
FNLPLGNYKK PKLLYCSNGG HFLRILPDGT VDGTRDRSDQ HIQLQLSAES AGEVYIKGTE TGQYLAMDTE GLLYGSQTPN EECLFLERLE ENHYNTYTSK KHAEKNWFVG LKKNGSCKRG PRTHYGQKAI LFLPLPVSSD
Greater than 90% as determined by SDS-PAGE.
14164
1 mg
Not determined.
Full Length
Mouse
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers