missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse AK1 Protein

A cDNA sequence encoding the AK1 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP10509-1ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15954088

  • 131.00 € / 1µg
Estimated Shipment: 26-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

AK1 Protein
Research Use Only
Specific activity is > 150 units/mg. One unit will convert 2.0 umoles of ADP to ATP + AMP per minute at pH 7.5 at 37C.
Mouse
His
AK1 protein solution (0.5 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 10% glycerol and 1mM DTT.
1 μg
< 1.0 EU per ug protein as determined by the LAL method.
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMGCCVSS EPQEEGGRKT GEKLKKAKII FVVGGPGSGK GTQCEKIVQK YGYTHLSTGD LLRAEVSSGS ERGKKLSAIM EKGELVPLDT VLDMLRDAML AKVDSSNGFL IDGYPREVKQ GEEFEQKIGQ PTLLLYVDAG AETMTQRLLK RGETSGRVDDNEETIKKRLE TYYNATEPVI SFYDKRGIVR KVNAEGTVDT VFSEVCTYLD SLK
Greater than 90.0% as determined by SDS-PAGE.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers