missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse AK1 Protein
A cDNA sequence encoding the AK1 was constructed and used to recombinantly synthesize the protein.
131.00 € - 1740.00 €
Specifications
Name | AK1 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | < 1.0 EU per ug protein as determined by the LAL method. |
Biological Activity | Specific activity is > 150 units/mg. One unit will convert 2.0 umoles of ADP to ATP + AMP per minute at pH 7.5 at 37C. |
Product Type | Recombinant Protein |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15954088
|
enQuireBio™
QP10509-1UG |
1 μg |
131.00 €
1µg |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15974088
|
enQuireBio™
QP10509-5UG |
5 μg |
203.00 €
5µg |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15964088
|
enQuireBio™
QP10509-50UG |
50 μg |
1740.00 €
50µg |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
AK1 Protein | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMGCCVSS EPQEEGGRKT GEKLKKAKII FVVGGPGSGK GTQCEKIVQK YGYTHLSTGD LLRAEVSSGS ERGKKLSAIM EKGELVPLDT VLDMLRDAML AKVDSSNGFL IDGYPREVKQ GEEFEQKIGQ PTLLLYVDAG AETMTQRLLK RGETSGRVDDNEETIKKRLE TYYNATEPVI SFYDKRGIVR KVNAEGTVDT VFSEVCTYLD SLK | |
Greater than 90.0% as determined by SDS-PAGE. |
Research Use Only | |
Specific activity is > 150 units/mg. One unit will convert 2.0 umoles of ADP to ATP + AMP per minute at pH 7.5 at 37C. | |
Mouse | |
His | |
AK1 protein solution (0.5 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 10% glycerol and 1mM DTT. |