missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Mouse AK2 Protein

Product Code. p-7145953 Shop All Bio Techne Products
Click to view available options
Quantity:
20 μg
5 μg
Unit Size:
20µg
5µg
This item is not returnable. View return policy

Product Code. 18286754

Brand: Novus Biologicals™ NBP26120120UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Highly purified and high bioactivity. Generating reliable and reproducible results. Applications: Functional, SDS-Page, Bioactivity

A bioactive recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-239 of Mouse AK2 The Recombinant Mouse AK2 Protein is derived from E. coli. The Recombinant Mouse AK2 Protein has been validated for the following applications: Functional, SDS-Page, Bioactivity.
TRUSTED_SUSTAINABILITY

Specifications

Concentration 0.5 mg/ml
For Use With (Application) Bioactivity, Functional Study, SDS-PAGE
Formulation PBS (pH 7.4) containing 10% glycerol.
Gene ID (Entrez) 204
Molecular Weight (g/mol) 29 kDa
Quantity 20 μg
Source E. coli
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Regulatory Status RUO
Gene Symbol AK2
Cross Reactivity AK2
Species Mouse
Purity or Quality Grade >95%, by SDS-PAGE
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.