missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse ASGPR1 / ASGR1 Protein
A DNA sequence encoding the Mus musculus (Mouse) ASGPR1 / ASGR1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP5684-ye-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001278060.1 | |
ASGPR1 / ASGR1 Protein | |
Research Use Only | |
Asgr1 | |
Recombinant Protein | |
Yeast | |
QNSQLREDLL ALRQNFSNLT VSTEDQVKAL STQGSSVGRK MKLVESKLEK QQKDLTEDHS SLLLHVKQLV SDVRSLSCQM AAFRGNGSER TCCPINWVEY EGSCYWFSSS VRPWTEADKY CQLENAHLVV VTSRDEQNFL QRHMGPLNTW IGLTDQNGPW KWVDGTDYET GFQNWRPEQP DNWYGHGLGG GEDCAHFTTD GRWNDDVCRR PYRWVCETKL DKAN | |
Greater than 90% as determined by SDS-PAGE. |
11889 | |
100 μg | |
Not determined. | |
Extracellular Domain | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |