missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse CD20 / MS4A1 Protein

A DNA sequence encoding the Mus musculus (Mouse) CD20 / MS4A1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP9289-ye-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16038021

  • 2100.00 € / 500µg
Estimated Shipment: 21-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_031667.1
CD20 / MS4A1 Protein
Research Use Only
MS4A1
Recombinant Protein
Yeast
VIMSSLSLFA AISGIILSIM DILNMTLSHF LKMRRLELIQ TSKPYVDIYD CEPSNSSEKN SPSTQYCNSI QSVFLGILSA MLISAFFQKL VTAGIVENEW KRMCTRSKSN VVLLSAGEKN EQTIKMKEEI IELSGVSSQP KNEEEIEIIP VQEEEEEEAE INFPAPPQEQ ESLPVENEIA P
Greater than 90% as determined by SDS-PAGE.
12482
500 μg
Not determined.
Partial (see sequence information for more details).
Mouse
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers